[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Microbiological Testing](https://www.sigmaaldrich.com/US/en/applications/microbiological-testing)Pyrogen testing methods # Pyrogen testing methods ![Three types of pyrogens are drawn in blue, cyan, and magenta against a yellow background.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/microbiological-testing/pyrogen-testing.jpg "What is a pyrogen?") Pyrogen testing determines the quantity of pyrogens in parenteral pharmaceutical or biopharmaceutical products and is regulated by several standards from organizations such as the Food and Drug Administration (FDA), United States Pharmacopeia (USP), or European Pharmacopeia (EP). The sterility of a product does not imply that it is free of pyrogens. Therefore, drugs that are purported to be sterile must also be tested for pyrogens to prevent febrile reactions in patients.  Pyrogen contamination can occur during production or the administration of pharmaceuticals, biotherapeutics, and medical devices, but the presence of pyrogens can also be an inherent characteristic of the product, such as adjuvants in vaccines or synthetic lipopeptides. [Request information](https://www.sigmaaldrich.com/US/en/campaigns/pyrogen-testing) * * * ## Related Products Slide 1 of 8 1 of 2 [![PyroMAT® kit kit containing all reagents to perform the Monocyte Activation Test (MAT) for Pyrogen testing, including the IL-6 Standard (control) and IL-6 ELISA kit, suitable for pyrogen testing](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/356/680/f0b931f5-6c6c-4d32-8193-9fbcca88a859/640/f0b931f5-6c6c-4d32-8193-9fbcca88a859.jpg) \ Millipore \ PYR0MATKIT \ PyroMAT® kit](https://www.sigmaaldrich.com/US/en/product/mm/pyr0matkit) Quick View [![PyroMAT® cells pkg of 2 vials, Cryopreserved Mono-Mac-6 cells for use in the Monocyte Activation Test (MAT) for Pyrogen testing](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/239/705/1733ae89-25c0-4f43-a466-2835f63f9a50/640/1733ae89-25c0-4f43-a466-2835f63f9a50.jpg) \ Millipore \ PYR0MATCELLS \ PyroMAT® cells](https://www.sigmaaldrich.com/US/en/product/mm/pyr0matcells) Quick View [![PyroMAT® Interleukin IL-6 standard Interleukin-6 standard for use in the Monocyte Activation Test (MAT) for Pyrogen testing](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/384/769/00485965-a49c-4354-a853-46a2a56e3dd3/640/00485965-a49c-4354-a853-46a2a56e3dd3.jpg) \ Millipore \ PYR0MATIL6 \ PyroMAT® Interleukin IL-6 standard](https://www.sigmaaldrich.com/US/en/product/mm/pyr0matil6) Quick View [![HKSA Non-Endotoxin Pyrogen (NEP) Control for use with PyroMAT® System](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/382/424/12822fc0-0813-44c0-9d9f-b2f320e8d657/640/12822fc0-0813-44c0-9d9f-b2f320e8d657.jpg) \ Millipore \ MATHKSA \ HKSA Non-Endotoxin Pyrogen (NEP) Control](https://www.sigmaaldrich.com/US/en/product/mm/mathksa) Quick View [![FLAGELLIN Non-Endotoxin Pyrogen (NEP) Control for use with PyroMAT® System](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/382/424/12822fc0-0813-44c0-9d9f-b2f320e8d657/640/12822fc0-0813-44c0-9d9f-b2f320e8d657.jpg) \ Millipore \ MATFLAGELLIN \ FLAGELLIN Non-Endotoxin Pyrogen (NEP) Control](https://www.sigmaaldrich.com/US/en/product/mm/matflagellin) Quick View [![Reference Standard Endotoxin EDQM endotoxin standard for Monocyte Activation Test](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/108/058/8b182a8f-5411-4e12-b58a-ef0a87879277/640/8b182a8f-5411-4e12-b58a-ef0a87879277.jpg) \ Millipore \ 1.44161 \ Reference Standard Endotoxin](https://www.sigmaaldrich.com/US/en/product/mm/144161) Quick View [![PyroDetect Interleukin Standard Interleukin-1Beta standard for use in the Monocyte Activation Test (MAT) for Pyrogen testing](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/391/323/527db9b8-0726-432a-8dab-6f8a5a67179b/640/527db9b8-0726-432a-8dab-6f8a5a67179b.jpg) \ Millipore \ 1.44158 \ PyroDetect Interleukin Standard](https://www.sigmaaldrich.com/US/en/product/mm/144158) Quick View [![Resiquimod Non-Endotoxin Pyrogen (NEP) Control for use with PyroMAT®](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/333/557/5529eb72-dcd0-42e2-b1eb-d0435966c966/640/5529eb72-dcd0-42e2-b1eb-d0435966c966.jpg) \ Millipore \ MATRESIQUIM0D \ Resiquimod Non-Endotoxin Pyrogen (NEP) Control](https://www.sigmaaldrich.com/US/en/product/mm/matresiquim0d) Quick View * * * ## Featured categories [![Open white box labeled PyroMAT Kit Pyrogen Detection Assay, with small bottles, vials, and a 96-well plate set of reagents arranged around it.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/pyromat-kit.png "PyroMAT® system")](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/pyrogen-testing) [Pyrogen testing](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/pyrogen-testing) Ensure safety: Pyrogen testing of pharmaceuticals with the Monocyte Activation Test (MAT) to detect endotoxin and non-endotoxin pyrogens. [Shop products](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/pyrogen-testing) [![A sophisticated sterility testing laboratory scene centered on a digital instrument with tubing. Surrounding it are various liquid-filled bottles in clear and amber tones, complemented by petri dishes with blue, red, and transparent media. A rack of red-capped vials suggests prepared samples or culture media. ](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/steritest-product-family.jpg "Sterility Testing Consumables, Media & Instruments")](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/sterility-testing-consumables-media-and-filtration) [Sterility testing consumables, media and instruments](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/sterility-testing-consumables-media-and-filtration) Laboratory instruments and consumables for sterility testing in microbiological quality control: Sterility test media, pumps, hardware and accessories. [Shop products](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/sterility-testing-consumables-media-and-filtration) Overview Articles and protocols Support ![A scientist wearing full PPE observes a sample on a microscope.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/microbiological-testing/pyrogen-testing-methods-application-page.jpg "Pyrogen Testing Methods - What is a Pyrogen?") ## [](https://www.sigmaaldrich.com)What is a pyrogen? A pyrogen is a substance that causes a rise in temperature (fever reaction) in a human or animal through the activation of the innate immune system. They constitute a heterogeneous group of contaminants comprising microbial and non-microbial substances. Pyrogens can be classified into two groups: endotoxins and non-endotoxin pyrogens (NEPs). Endotoxins are substances found in Gram-negative bacteria. Non-endotoxin pyrogens are other microbial substances, including those derived from Gram-positive bacteria, viruses, yeasts and fungi. Non-microbial pyrogenic substances can also originate from rubber particles, microscopic plastic particles, or metal compounds in elastomers. Several test methods are available for the detection of pyrogens. They can be classified based on the type of contaminant they detect, and the need for animal materials to perform the test as described in the below table: | | | | |---------------------|-----------------------------------------------------|--------------------------------| | | Detection of endotoxins and non-endotoxins pyrogens | Detection of endotoxins only | | Animal-based method | Rabbit pyrogen test (RPT) | Bacterial endotoxin test (BET) | | *In vitro* method | Monocyte activation test (MAT) | Recombinant BET (r-FC, r-CR) | ## [](https://www.sigmaaldrich.com)Rabbit pyrogen testing The rabbit pyrogen test (RPT) involves measuring the rise in temperature of rabbits following the intravenous injection of the tested product. The RPT gives qualitative results and the sensitivity is quite low. The robustness of the test is also limited, due to development of pyrogen tolerance in rabbits after repeated injections, or stress from the rabbits when performing the assay. Note: Rabbit Pyrogen Testing (RPT) is now forbidden in Europe, and the Monocyte Activation Test (MAT) is the method of choice for detecting both endotoxin and non-endotoxin pyrogens. ## [](https://www.sigmaaldrich.com)Monocyte Activation Test The [Monocyte Activation Test (MAT)](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/pyrogen-testing) is an alternative to animal-based methods for the detection of both endotoxin and non-endotoxin pyrogens. The monocyte activation test mimics the human immune reaction by incubating monocytes with the tested sample. If pyrogens are present, monocytes are activated and produce inflammatory molecules, cytokines, responsible for the febrile reaction. The cytokines are then detected using an immunological assay (ELISA) involving specific antibodies and an enzymatic color reaction.  Note: As of July 2025, the European Pharmacopoeia (Ph. Eur.) Commission recommends using the Monocyte Activation Test (MAT) as the preferred method for endotoxin and non-endotoxin pyrogen testing. The Rabbit Pyrogen Test (RPT) is now banned in Europe. [Read the article](https://www.edqm.eu/en/-/ph.-eur.-bids-adieu-to-rabbit-pyrogen-test-in-its-monographs) and [discuss further with our Experts](https://www.sigmaaldrich.com/US/en/campaigns/pyrogen-testing) to move to the MAT detecting endotoxin and non-endotoxin pyrogens and to follow the regulatory trend.  ## [](https://www.sigmaaldrich.com)Bacterial endotoxin testing (LAL test) The most common method for endotoxin testing is the Limulus amoebocyte lysate test (LAL test); an assay based on the lysate of amoebocytes from the horseshoe crab blood. The lysate from horseshoe crab blood cells naturally reacts with bacterial endotoxins in a coagulation reaction. This method has a high sensitivity for the quantification of endotoxins, but it does not detect non-endotoxin pyrogens. ## [](https://www.sigmaaldrich.com)Recombinant BET (r-FC, r-CR) testing The recombinant Factor C is a genetically engineered protein normally found in the Limulus amebocyte lysate cascade. In this test, Factor C reacts with endotoxin and couples with a marker to produce a quantifiable, fluorescent end product. The recombinant Factor C test uses the same principle as the LAL test, but without the need for animal-derived material.  The recombinant Cascade Reagent (r-CR) is a genetically engineered multi-component reagent that reconstitutes the complete coagulation cascade found in the Limulus amebocyte lysate. In this test, the cascade proteins,Factor C, Factor B, and the proclotting enzyme, react sequentially with endotoxin to produce a quantifiable end product. The r-CR test replicates the full LAL pathway, but without the need for animal-derived material. [Shop all products](https://www.sigmaaldrich.com/search/pyrogen-testing?focus=products&page=1&perpage=30&sort=relevance&term=Pyrogen%20Testing&type=product) [![Document search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png "Document search")](https://www.sigmaaldrich.com/documents-search) [Looking for more specific information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find documents](https://www.sigmaaldrich.com/documents-search) * * * ## Related articles - [How to Choose the Right Method for Microbial Testing in Infant Formula](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/infant-formula-interview) Interview: Accurate microbiology QC methods for infant formula testing - [Probiotics and Human Health](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/probiotics-and-human) Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system. - [Assessing Level of Bacteria Purified Water](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/assessing-level-of-bacteria-purified-water) Best practices for microbial testing of purified water in the lab. Covers storage, culture and incubation conditions, and contaminants reduction. - [Base Ingredients of Microbiology Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/microbiological-testing/microbial-culture-media-preparation/base-ingredients-of-microbiology-media) Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators. - [MC-Media Pad – Validation Summary](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/microbiological-testing/microbial-culture-media-preparation/mc-media-pad-validation-summary) This article will focus on results obtained for MC-Media Pad RAC, MC-Media Pad Coliform, MC-Media Pad *E. coli* & Coliform & MC-Media Pad Yeast & Mold. - [See all (9)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related protocols - [Enterobacteriaceae in Food](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/enterobacteriaceae-in-food) General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively. - [See all (1)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) ### Find more articles and protocols Enter Keywords Search * * * ## Related webinars [![Webinar Monocyte Activation Test](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/video-posters/monocyte-activation-test-poster.jpg "Webinar Monocyte Activation Test")](https://www.sigmaaldrich.com/US/en/collections/webinars/w924713416) [Monocyte activation test: A powerful tool to assess pyrogenic risk in pharmaceutical process](https://www.sigmaaldrich.com/US/en/collections/webinars/w924713416) In this webinar, we discuss how monocyte activation tests performed with the PyroMAT® System detect endotoxin and non-endotoxin pyrogens. [![Benefits of the PyroMAT® system and how to use it](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/video-posters/pyromat-system-benefits-poster.jpg "Benefits of the PyroMAT® system and how to use it")](https://www.sigmaaldrich.com/US/en/collections/webinars/w877364779) [*In Vitro* Pyrogen detection in pharma](https://www.sigmaaldrich.com/US/en/collections/webinars/w877364779) Learn how our PyroMAT® System provides a robust solution for in vitro Pyrogen Test in pharma with a ready-to use kit. [![Risk considerations for the presence of pyrogens and the need for the MAT test in pharmaceutical processing](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/video-posters/mat-test-in-pharmaceutical-processing-webinar-poster.jpg "Risk considerations for the presence of pyrogens and the need for the MAT test in pharmaceutical processing")](https://www.sigmaaldrich.com/US/en/collections/webinars/w343863313) [Risk considerations for the presence of pyrogens and the need for the MAT test in pharmaceutical processing](https://www.sigmaaldrich.com/US/en/collections/webinars/w343863313) In this lecture, you will learn how to test for pyrogen (including non-endotoxin pyrogens) in your pharmaceutical samples, what are the existing methods in order to have a controlled process. ## How can we help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional support - [Chromatogram search](https://www.sigmaaldrich.com/chromatogram-search) Use the Chromatogram search to identify unknown compounds in your sample. - [Calculators and apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our frequently asked questions for answers to commonly asked questions about our products and services. Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map) An unknown error has occured.