Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Solid Phase Extraction
SPE Method Development
Reversed-phase SPE
Ion-exchange SPE
Normal phase SPE
FREE SPE Sample Packs for Method Development
Supelclean™ SPE Method Development Kits
96-Well SPE Method Development Plate
Application Assistance
Request Method Development
Poster
FREE SPE Sample Packs for Method Development back to top
SPE Sample Packs consist of an assortment of SPE phase chemistries and tube dimensions ideally suited for method development. The mix of phase chemistries available in these Sample Packs allows you to screen for optimal retention and selectivity required to achieve your sample prep objectives.
To view available phases and request a sample, visit our SPE Sample Packs page.
You can also contact Technical Service at 814-359-3041 (800-359-3041 within the U.S.) or techservice@sial.com.
View available SPE phases and request a sample.
Supelclean SPE Method Development Kits back to top
Our kits consist of an assortment of SPE phase chemistries and cartridge configurations ideal for SPE method development. The range of phase chemistries available for each kit allows the user to profile for compund retention, elution and sample matrix selectivity.
Supelclean SPE Method Development Kit Kit A Kit B Kit C Kit NP-3 Kit IX-3
Supelclean Packing Sorbent Qty. / Tube Size
LC-Si 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL
1 g/6 mL
500 mg/3 mL
LC-8 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL
LC-18 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL
LC-CN 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL 500 mg/3 mL
LC-Diol 500 mg/3 mL 100 mg/1 mL 500 mg/3 mL
LC-NH2 500 mg/3 mL 100 mg/1 mL 500 mg/3 mL 500 mg/3 mL
LC-Ph 500 mg/3 mL 100 mg/1 mL
LC-SAC 500 mg/3 mL 100 mg/1 mL 500 mg/3 mL
LC-SCX 500 mg/3 mL 100 mg/1 mL 500 mg/3 mL
LC-WCX 500 mg/3 mL 100 mg/1 mL 500 mg/3 mL
LC-Alumina-A 1 g/6 mL 1 g/3 mL
LC-Alumina-B 1 g/6 mL 1 g/3 mL
LC-Alumina-N 1 g/6 mL 1 g/3 mL
LC-Florisil 1 g/6 mL
Quantity each tube 6 12 3 6 12
Catalog Number 57019 57009-U 57075-U 57074-U 57073
96-Well SPE Method Development Plates back to top
 Supelco 96-well SPE Method Development Plates contain an assortment of SPE phase chemistries ideally suited for method development. The mix of phase chemistries contained within this 96-well SPE plate allows researchers to screen for analyte retention, recovery, and selectivity when achieving one’s sample prep objectives.
For ordering information, go to ourOn-Line Catalog
Additional Resources
Subject No.
Pages
File Size Id.
Improve Sample Prep Selectivity through 96-well SPE Method Development Plates 3 311 Kb T206115
Application Assistance back to top
For application assistance, please use the Application Assistance Form to provide your requirements. A technical service representative will contact you to assist you in developing/troubleshooting your sample prep method.