Spectroscopy
NMR Spectroscopy Tubes
NMR Solvents / NMR Tubes
Spectral Viewer / Synthesis
Spectroscopy Home
NMR Tubes
NMR Tubes / Wilmad NMR Tubes / Wilmad Screw Cap NMR Tubes
Norell™ NMR Sample Tubes
/ Shigemi Advanced NMR microtube assembly / Cross-Reference Guide
NMR Tubes
Every premium-quality NMR tube is inspected and gauged to meet published specifications.
  • Statement of Conformance stamped on each box of tubes
  • Made of ASTM Type I borosilicate glass* for compatibility with ground joints, valves, and vacuum racks
  • Precision ground and highly polished for spinning stability and highest resolution
  • Thin-wall glass with round-bottoms
  • Sandblasted marking spot for labelling samples
    *Except disposable grade NMR tubes.
NMR TUBE SELECTION GUIDE
Choose the NMR tube grade that most closely matches the frequency (in MHz) of the spectrometer.
Grade MHz Applications Cross Ref. No.
(5mm tubes)
Wilmad Kontes
Disposable 30 Student labs, basic studies WG-Economy
Series 30 30 Student labs, basic studies 505-PS 897200
Series 200 60 to 270 Lower field studies 506-PP 897205
Series 300 60 to 360 Routine applications 507-PP 897220
Series 400 200 to 500 High-field, multiple-scan FT-NMR 528-PP 897235
Series 500 500+ Ultra-high resolution 535-PP 897240
Series 800 600 to 800 Highest resolution and performance 541-PP 897245
Grade L (in.) Wall uniformity
(in., TIR)
Camber (in.) Cat. No.
Add to Cart
3mm Microsample NMR tubes — 0.012in. wall thickness (Pkg=5 tubes)
Series 300 7 0.0020 0.0010 Z412902
  8 0.0020 0.0011 Z412910
Series 400 7 0.0010 0.0005 Z412929
  8 0.0010 0.0006 Z412937
Series 500 7 0.0005 0.00025 Z412945
  8 0.0005 0.00035 Z412953
5mm NMR tubes — 0.015in. wall thickness (Pkg=50 tubes)
Series 60 7 - - Z543888
  8 - - Z555908
5mm Thrift NMR tubes - 0.01475in. wall thickness (pkg=5 tubes)
Series 200 7 0.002 0.0005 Z555894
  8 0.002 0.0006 Z556092
Series 300 7 0.0015 0.001 Z556106
  8 0.0015 0.001 Z556114
Series 400 7 0.0005 0.00025 Z556122
  8 0.0005 0.00035 Z556130
Series 500 7 0.0005 0.00025 Z556149
  8 0.0005 0.00035 Z556157
5mm Precision NMR tubes - 0.01475in. wall thickness (pkg=5 tubes)
Series 80 7 0.003 0.002 Z412783
Series 200 7 0.0025 0.002 Z412791
Series 300 7 0.002 0.005 Z412805
  8 0.002 0.0006 Z412813
  9 0.002 0.0007 Z412821
Series 400 7 0.001 0.0005 Z412848
  8 0.001 0.006 Z412856
Series 500 7 0.0005 0.00025 Z412864
  8 0.0005 0.00035 Z412872
Series 800 7 0.0002 0.00015 Z412880
  8 0.0002 0.00015 Z412899
Series 900 7 0.0002 0.00015 Z550868
  8 0.0002 0.00015 Z548243
Wilmad NMR Tubes
Made of Pyrex borosilicate glass (unless otherwise noted) for compatibility with ground joints, valves, and vacuum racks. Precision ground and highly polished for spinning stability and highest resolution. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes that are 9in. L are for making standards and for use with tip-off manifolds.
3mm Microsample NMR tubes - 0.012in. wall thickness (Pkg=5 tubes)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
300MHz tubes
7 51/25 307-PP-7 Z274321
8 51/25 307-PP-8 Z274348
9 51/25 307-PP-9 Z274356
360MHz tubes
7 25/25 327-PP-7 Z271861
8 25/25 327-PP-8 Z274364
9 25/25 327-PP-9 Z274372
400MHz tubes
7 25/13 328-PP-7 Z274380
8 25/13 328-PP-8 Z274399
9 25/13 328-PP-9 Z274402
500MHz tubes
7 13/6 335-PP-7 Z274410
8 13/6 335-PP-8 Z274429
9 13/6 335-PP-9 Z274437
Back to Top
5mm amber NMR tubes - 0.015in. wall thickness (Pkg=1 tube)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
100MHz tube
7 76/51 505-PS-7 Z282251
360MHz tubes
7 51/25 507-PP-7 Z282278
7 25/25 527-PP-7 Z282286
Back to Top
5mm disposable NMR tubes - 0.015in. wall thickness, N51 glass (Pkg=100 tubes)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
100MHz tubes
7 76/76 HIP-7 Z514144
8 76/76 HIP-8 Z514152
Back to Top
5mm economy NMR tubes - 0.015in. wall thickness. WG series made of N51 glass. (Pkg=5 tubes)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
100MHz tubes
7 76/76 WG-5-7 Z276278
8 76/76 WG-5-8 Z276286
9 76/76 WG-5-9 Z276294
7 76/76 512-5-7 Z276308
8 76/76 512-5-8 Z276316
9 76/76 512-5-9 Z276324
7 51/51 506-IM-7 Z514128
8 51/51 506-IM-8 Z514136
Back to Top
5mm standard NMR tubes - 0.015in. wall thickness (Pkg=5 tubes)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
100MHz tubes
7 76/51 505-PS-7 Z271985
8 76/51 505-PS-8 Z274445
9 76/51 505-PS-9 Z274453
7 51/51 506-PP-7 Z286028
8 51/51 506-PP-8 Z286036
9 51/51 506-PP-9 Z286044
200MHz tube
7 76/51 503-PS-7** Z410357
360MHz tubes
7 51/25 507-PP-7 Z271993
8 51/25 507-PP-8 Z274461
9 51/25 507-PP-9 Z272116
7 51/13 526-PP-7 Z274488
8 51/13 526-PP-8 Z274496
9 51/13 526-PP-9 Z274518
7 25/25 527-PP-7 Z274275
8 25/25 527-PP-8 Z274526
9 25/25 527-PP-9 Z274534
400MHz tubes
7 25/13 528-PP-7 Z272019
8 25/13 528-PP-8 Z274291
9 25/13 528-PP-9 Z274542
500 MHz tubes
7 13/6 535-PP-7 Z272027
7 13/6 545-PPT-7* Z406708
8 13/6 535-PP-8 Z274550
9 13/6 535-PP-9 Z274569
800 MHz tubes
7 3.8/3.8 541-PP-7 Z412007
8 3.8/3.8 541-PP-8 Z412015
900 MHz tubes
7 2.5/3.8 542-PP-7 Z412023
8 2.5/3.8 542-PP-8 Z412031
* Ultra thin wall (0.09in.)
** Medium wall (0.03in.)
10mm NMR tubes - 0.018in. wall thickness (Pkg=5 tubes)
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
60MHz tubes
7 NA 513-1PS-7 Z274771
8 NA 513-1PS-8 Z274798
9 NA 513-1PS-9 Z274801
100MHz tubes
7 254/51 Z27,468-2 Z272035
8 254/51 513-1PP-8 Z274690
9 254/51 513-1PP-9 Z274704
300MHz tubes
7 76/38 513-3PP-7 Z272035
8 76/38 513-3PP-8 Z274712
9 76/38 513-3PP-9 Z272043
360MHz tubes
7 51/25 513-5PP-7 Z274720
8 51/25 513-5PP-8 Z274739
9 51/25 513-5PP-9 Z274747
400MHz tubes
7 38/13 513-7PP-7 Z272051
8 38/13 513-7PP-8 Z274755
9 38/13 513-7PP-9 Z274763
*Tubes do not have a precision i.d. tolerance.
Wilmad Screw Cap NMR Tubes
NMR tube has an open-top, screw-thread cap with a PTFE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable. Excellent for handling air-sensitive samples and in cell perfusion studies. Tubes are 7in. L.
5mm NMR tubes  (Pkg=1 tube)
Product Name Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
100MHz tube 76/51 505-TR-7 Z271918
100MHz tube 51/51 506-TR-7 Z271926
360MHz tube 51/25 507-TR-7 Z271934
400MHz tube 25/13 528-TR-7 Z271942
360MHz tube 51/25 507-TR-7 Z271934
400MHz tube 25/13 528-TR-7 Z271942
500MHz tube 13/60 535-TR-7 Z271950
8mm NMR tubes  (Pkg=1 tube)
Product Name Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
400MHz tube 38/13 513A-7TR Z274828
10mm NMR tubes  (Pkg=1 tube)
Product Name Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
300MHz tube 76/38 513-3TRA-7 Z271969
400MHz tube 38/13 513-7TRA-7 Z271977
Replacement caps and septa for screw-cap NMR tubes
NMR tube size (mm) Cat. No. Pkg
Add to Cart
Black open-top caps
5 Z162531 144
10 Z106453 200
Black solid-top caps w/white rubber liner
5 Z188808 100
10 Z106410 200
Black solid-top caps w/PTFE-faced rubber liner
10 Z106437 200
PTFE-faced silicone septa
5 Z162566 120
10 Z106518 100
Norell™ NMR Sample Tubes
MHz
Length (in.)
Concentricity +/-(mm)
Camber +/- (mm)
Cat. No.
Add to Cart
3 mm Select Series™ - ASTM Type 1 Class A Glass
o.d. (mm) 2.99 (+/- 0.004)
i.d. (mm) 2.41 (+/- 0.006)
900
7
0.002
0.003
900
8
0.002
0.003
800
7
0.025
0.0038
800
8
0.025
0.0038
600
7
0.004
0.006
600
8
0.004
0.006
500
7
0.005
0.013
500
8
0.005
0.013
400
7
0.007
0.019
400
8
0.007
0.019
300
7
0.007
0.025
300
8
0.007
0.025
200
7
0.01
0.038
200
8
0.01
0.038
5 mm Select Series™ - ASTM Type 1 Class A Glass
o.d. (mm) 4.97 (+/- 0.004)
i.d. (mm) 4.20 (+/- 0.006)
900
7
0.002
0.003
900
8
0.002
0.003
800
7
0.0025
0.0038
800
8
0.0025
0.0038
600
7
0.004
0.006
600
8
0.004
0.006
500
7
0.005
0.013
500
8
0.005
0.013
400
7
0.007
0.019
400
8
0.007
0.019
300
7
0.007
0.025
300
8
0.007
0.025
200
7
0.009
0.035
200
8
0.009
0.035
5 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass
o.d. (mm) 4.97 (+/- 0.013)
i.d. (mm) 4.20 (+/- 0.025)
600
7
0.004
0.006
600
8
0.004
0.006
500
7
0.005
0.013
500
8
0.005
0.013
400
7
0.007
0.019
400
8
0.007
0.019
300+
7
0.007
0.025
300+
8
0.007
0.025
300
7
0.01
0.038
300
8
0.01
0.038
200
7
0.01
0.04
200
8
0.01
0.04
100
7
0.02
0.07
100
8
0.02
0.07
60
7
nominal
nominal
60
8
nominal
nominal
10 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass
o.d. (mm) 10.00 (+/- 0.013)
i.d. (mm) 8.76 (+/- 0.025)
400
7
0.005
0.007
400
8
0.005
0.007
300
7
0.02
0.013
300
8
0.02
0.013
200
7
nominal
nominal
200
8
nominal
nominal
Back to Top
Shigemi Advanced NMR microtube assembly
?Outer Sample Tube O.D. (mm)
Bottom L (mm)
Shigemi No.
Cat. No.
Add to Cart
Matched with CDCl3
5
8
CMS-005TB
5
12
CMS-005TJ
5
15
CMS-005TV
8
8
CMS-008TB
8
12
CMS-008TJ
8
15
CMS-008TV
Matched with CD3OD
5
8
MMS-005TB
5
12
MMS-005TJ
5
15
MMS-005TV
8
8
MMS-008TB
8
12
MMS-008TJ
8
15
MMS-005TV
Matched with DMSO-d6
5
8
DMS-005TB
5
12
DMS-005TJ
8
8
DMS-008TB
8
12
DMS-008TJ
8
15
DMS-008TV
5
15
DMS-005TV
Matched with D2O
5
8
BMS-005TB
5
12
BMS-005TJ
5
15
BMS-005TV
8
8
BMS-008TB
8
12
BMS-008TJ
8
15
BMS-008TV
Matched with CDCl3
8
-
CMS005B
12
-
CMS005J
15
-
CMS005V
-
Microtube cap
CMSCAP
Matched with DMSO-d6
8
-
DMS005B
12
-
DMS005J
15
-
DMS005V
-
Microtube cap
DMSCAP
Matched with D2O
8
-
BMS005B
12
-
BMS005J
15
-
BMS005V
-
Microtube cap
BMSCAP
Cross-Reference Guide
?Norell™
Wilmad®
     Prod #
Norell™
Wilmad®
       Prod #
NORST5007
HIP-7
NORS36007
335-PP-7
NORST5008
HIP-8
NORS36008
335-PP-8
NORST5007
WG-5MM-ECON-7
NORXR557
505-PS-7
NORST5008
WG-5MM-ECON-8
NORXR558
505-PS-8
NOR5027
512-7
NORS52007
506-PP-7
NOR5028
512-8
NORS52008
506-PP-8
NOR5027
1206-7
NORS53007
507-PP-7
NOR5028
1206-8
NORS53008
507-PP-8
NOR505P7
1208-07
507-PP-9
NOR505P8
1208-08
NOR10017
513-1PP-7
NORXR557
1226-7
NOR10018
513-1PP-8
NORXR558
1226-8
NOR1005P7
513-3PP-7
NOR506P7
1228-7
NOR1005P8
513-3PP-8
NOR506P8
1228-8
NOR1008UP7
513-7PP-7
NOR507HP7
1235-7
NOR1008UP8
513-7PP-8
NOR507HP8
1235-8
NORS54007
526-PP-7
NOR508UP7
1241-7
NORS54008
526-PP-8
NOR508UP8
1241-8
NORS55007
528-PP-7
NOR509UP7
1242-7
NORS55008
528-PP-8
NOR509UP8
1242-8
NORS56007
535-PP-7
NORS33007
305-PS-7
NORS56008
535-PP-8
NORS33008
305-PS-8
NORS58007
541-PP-7
NORS34007
307-PP-7
NORS58008
541-PP-8
NORS34008
307-PP-8
NORS59007
542-PP-7
NORS35007
328-PP-7
NORS59008
542-PP-8
NORS35008
328-PP-8