Titration
Aquastar® Karl Fischer Reagents
Aquastar® reagents for Karl Fischer titration enable determination of water content in gaseous, liquid and solid samples with high accuracy and reliability. Our high-quality Aquastar® product portfolio comprises a whole range of Karl Fischer reagents for volumetric and coulometric determinations. In addition, for monitoring and qualifying Karl Fischer instruments, checking measuring results and determining the titer of the reagents, we offer various types of water standards.
Resources and References