Innovative Packaging - Optimized for Every Application
In order to ensure highest quality for our products, optimized packaging is needed to protect the reagents from impurities and contamination. We offer a wide range of high-quality packaging options for volumetric solutions, using glass and plastic grades that do not alter any of the product characteristics. All our packaging materials are tested for their quality and atmospheric permeability, thereby ensuring the purity of the solutions, also during storage up to their minimum shelf life for sealed, original containers.
Titripur® - ready-to-use solutions reliable and precise
We offer ready-to-use volumetric solutions for daily laboratory analyses. These can be connected to a titrator via an adapter. Depending on the amount required, volumetric solutions are available in 0.5L, 1L, 2.5L, 5L, 10L and 25L PE bottles or canisters. In addition, volumetric solutions which cannot be stored in a stable manner for an extended period of time in PE containers are available in 1L and 2.5L glass bottles. Also Titripur grade ready-to-use solutions are offered in 4L and 10L Titripac containers.
Polyethylene bottles and canisters:
  • Diversity: Packaging material for all requirements
  • Unbreakable and contamination-free material
  • Direct connection to titrator
  • Stability of reagents ensured for entire shelf life
Glass bottles:
  • Ensuring chemical stability of special volumetric solutions
Titripac® - reliable solution from the first to the last drop
Our Titripac is an innovative and safe packaging option for high-quality ready-to-use volumetric solutions. Its economic and ecological advantages enable you to optimize your workflow. The consistent quality of a volumetric solution is ensured from the first to the last drop. This is enabled by an hermetically sealed packaging system: contamination caused by air, carbon dioxide or microorganisms is inhibited. Titripac saves you the time-consuming process of checking the solutions as well as the cost-intensive disposal of contaminated residual amounts. The carton can simply be disposed of together with recovered paper. The internal liner can be easily folded together prior to disposal to minimize waste volume.
Titripac is extremely easy to use. The integrated spout appears by simply pressing on the pack. After opening the tap, liquid can be withdrawn at any time - conveniently and without risk of contamination. In addition, Titripac can be connected directly to the titrator via an adapter.
  • No contamination: Hermetically sealed pack
  • Easy to use: Integrated withdrawal tap, direct connection to titrator possible
  • Saves time and money: No unnecessary titer determinations, no contaminated residual amounts
  • Environmentally friendly disposal: Reduced packaging waste, carton and internal liner can be disposed of separately
Titrisol® - concentrates for greater flexibility
For users who regularly perform different types of analyses, we also offer most volumetric solutions as Titrisol concentrates. Every ampoule contains a precisely defined quantity of substance that is normally filled to a final volume of 1 liter. However, any other concentrations can be produced by diluting correspondingly.
  • Space-saving
  • Flexible and easy to use
  • Individual concentrations can be prepared