[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/CA/en) Products Cart0 CAEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/CA/en)[Applications](https://www.sigmaaldrich.com/CA/en/applications)[Analytical Chemistry](https://www.sigmaaldrich.com/CA/en/applications/analytical-chemistry)[Sample Preparation Techniques](https://www.sigmaaldrich.com/CA/en/applications/analytical-chemistry/sample-preparation)QuEChERS Sample Preparation Method # QuEChERS Sample Preparation Method ![Various sorbents exist for the QuEChERS cleanup step, chosen according to the matrix of interest.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/analytical-chemistry/quechers-tubes-card.jpg "Various sorbents exist for the QuEChERS cleanup step, chosen according to the matrix of interest.") QuEChERS is a type of dispersive solid phase extraction (dSPE) used for sample preparation. QuEChERS stands for ‘Quick, Easy, Cheap, Effective, Rugged and Safe’. Originally developed to prepare fruit and vegetable samples for pesticide residue analysis, this method is now used to analyze other chemical classes, including PAHs (polycyclic aromatic hydrocarbons), PCBs (polychlorinated biphenyls), PBDEs (polybrominated diphenyl ethers), and flame retardants. The concept was introduced in 2002 and is now specified in official methods of the Association of Analytical Communities (AOAC 2007.01) and European Committee for Standardization (EN 15662:2008). * * * ## Related Products Slide 1 of 11 1 of 3 [![Supel™ QuE Citrate/Sodium Bicarbonate Tube, pk of 50](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/645/638/63abfc2a-28ed-4f1b-8e9f-a5b67e0d7684/640/63abfc2a-28ed-4f1b-8e9f-a5b67e0d7684.jpg) \ Supelco \ 55237-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55237u) Quick View [![Supel™ QuE Acetate Tube, pk of 50, AOAC® 2007.01](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/125/286/44d207e0-932f-40bf-98c2-535130337ac5/640/44d207e0-932f-40bf-98c2-535130337ac5.jpg) \ Supelco \ 55234-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55234u) Quick View [![Supel™ QuE Non-Buffered Tube 1, pk of 50](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/228/944/c292b893-9107-4977-a204-15c70a3081f8/640/c292b893-9107-4977-a204-15c70a3081f8.jpg) \ Supelco \ 55294-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55294u) Quick View [![QuEChERS Shaker and Rack Starter Kit AC/DC input 115 V AC (USA Compatible Plug)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/768/587/4c9ebceb-9071-47fe-95a3-7e47e25ca4ff/640/4c9ebceb-9071-47fe-95a3-7e47e25ca4ff.jpg) \ Supelco \ 55278-U \ QuEChERS Shaker and Rack Starter Kit](https://www.sigmaaldrich.com/CA/en/product/supelco/55278u) Quick View [![50 mL QuEChERS Extraction Tube Shaker Rack Horizontal foam shaker rack, holds 7 extraction tubes](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/425/135/e1b03e19-eb43-404c-ada3-8f9216b1ca40/640/e1b03e19-eb43-404c-ada3-8f9216b1ca40.jpg) \ Supelco \ 55279-U \ 50 mL QuEChERS Extraction Tube Shaker Rack](https://www.sigmaaldrich.com/CA/en/product/supelco/55279u) Quick View [![Supel™ QuE PSA Tube, pack of 50 ea, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55437-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55437u) Quick View [![Supel™ QuE PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55464-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55464u) Quick View [![Supel™ QuE PSA/C18 Tube (Shaker Compatible), pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55439-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55439u) Quick View [![Supel™ QuE PSA Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55466-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55466u) Quick View [![Supel™ QuE PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55474-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55474u) Quick View [![Supel™ QuE PSA/C18 Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/214/757/3d923f6d-cfc9-495e-bddb-d524abd91075/640/3d923f6d-cfc9-495e-bddb-d524abd91075.jpg) \ Supelco \ 55470-U \ Supel™ QuE](https://www.sigmaaldrich.com/CA/en/product/supelco/55470u) Quick View * * * ## Featured Categories [![A laboratory device featuring a clear chamber with five vertical syringes on top, connected to a pressure gauge and a valve at the base. The device is branded with the logo 'Supelco' and vibrant M logo on a bright yellow background.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/analytical-sample-preparation/spe-12-port-vacuum-manifold.jpg)](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers) [Solid Phase Extraction (SPE) and QuEChERS](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers) Discover our range of solid phase extraction products for rapid and accurate analytical sample preparation to ensure effective and reliable chromatographic analysis. They include SPE cartridges, plates, extraction tubes and dispersive SPE tools, and accessories [Shop Products](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers) [![Transparent coiled GC column with metal end fittings against a white background, essential for separating mixtures in gas chromatography. A GC capillary column with a stationary phase.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/analytical-chemistry/gc-columns-green-background-400-248.jpg)](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/gc-columns-accessories-and-adsorbents) [GC Columns, Accessories & Adsorbents](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/gc-columns-accessories-and-adsorbents) Achieve superior GC performance with our columns, consumables, and accessories. Explore Supelco® offerings. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/gc-columns-accessories-and-adsorbents) [![3 Bottles of ReagentPlus® solvent grade products in various volumes.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/analytical-chromatography/reagentplus-solvent-grade-products.jpg)](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/solvents) [Solvents](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/solvents) Your Solvent Source: Find the right fit with Supelco®, SigmaAldrich®, and SAFC® brands, covering analytical, lab, and biopharmaceutical uses. Order online. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/solvents) [![A collection of various Supelco® HPLC and UHPLC columns scattered on a green surface. These columns, which are cylindrical with metallic finishes and equipped with fittings at both ends, represent a range of chromatography column technologies for different analytical needs. They vary in size and some have labeling that indicates specifications or model numbers.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/analytical-chromatography/high-performance-liquid-chromatography-products.jpg)](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/hplc-columns) [HPLC Columns](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/hplc-columns) Achieve precise separations with our extensive HPLC column collection. Enhance retention, resolution, and selectivity. Order today. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/analytical-chemistry/analytical-chromatography/hplc-columns) Overview Related Articles & Protocols Support ## [](https://www.sigmaaldrich.com)QuEChERS Sample Extraction Solvent extraction techniques are designed to achieve maximum analyte yield from the base matrix. Analytes of interest are extracted from the sample matrix using water-miscible solvents (usually acetonitrile) and high concentrations of salt and/or buffering agents. Solvent selection is important to minimize compound co-extraction. Salts are used in the extraction step to create phase separation. They act as buffers to adjust the pH and remove water from the sample.  ## [](https://www.sigmaaldrich.com)QuEChERS Cleanup Sample cleanup reduces interference during subsequent analysis, as interference can damage analytical instrumentation and complicate analyte identification and quantification. Unlike traditional sample cleanup methods using SPE tubes, in QuEChERS, cleanup is facilitated by mixing bulk amounts of SPE sorbents with the sample extract. Sorbents used in clean up remove matrix interferences that coextract with analytes of interest. The use of PSA, C18 or GCB (graphitized carbon black) sorbents remove sugars, lipids, sterols, organic acids, proteins, carotenoids, chlorophyll, and other pigments prior to a GC-MS/MS or LC-MS/MS analysis. Special Z-Sep sorbents may be needed to cleanup difficult matrices containing pigments or fat. [![Document Search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png)](https://www.sigmaaldrich.com/documents-search) [Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find Documents](https://www.sigmaaldrich.com/documents-search) ## Related Articles - [Comparison of QuEChERS Products for the Extraction of Planar Pesticides from Spinach](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/planar-pesticides-spinach) Supel™ QuE Verde combines a novel carbon with zirconia coated silica (Z-Sep+) to provide an optimum balance between planar pesticide recovery and color removal. - [Pesticide Residue Analysis in Ginger Powder with QuEChERS](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/analysis-of-pesticide-residues-in-ginger-powder) 2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis. - [Supel™ QuE Z-Sep Sorbents for QuEChERS](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/quechers) Z-Sep sorbents improve fat and pigment extraction in QuEChERs solid phase extraction for sample preparation. - [Analysis of Active Cannabis Compounds](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/analysis-of-active-cannabis-compounds) HPLC-UV analysis of active compounds in marijuana-infused edibles for potency testing; complex matrix requires precise methods. - [Analysis of Fipronil and Fipronil Sulfone in Eggs, Chicken Meat and Mayonnaise](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/analysis-of-fipronil-and-fipronil-sulfone) A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone. - [See All (9)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related Protocols - [Analysis of Active Cannabis Compounds](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/analysis-of-active-cannabis-compounds) HPLC-UV analysis of active compounds in marijuana-infused edibles for potency testing; complex matrix requires precise methods. - [Analysis of Fipronil and Fipronil Sulfone in Eggs, Chicken Meat and Mayonnaise](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/analysis-of-fipronil-and-fipronil-sulfone) A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone. - [QuEChERS Method for Pesticide Residue Analysis](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/quechers-method-pesticide-residue-analysis) QuEChERS method for pesticide residue analysis requires only small quantities of solvent and is capable of generating recoveries of 70-120% with RSDs <5% for a wide range of compounds. - [Neonicotinoids Ban: Impact on Plant Material](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/analytical-chemistry/quechers/banned-other-neonicotinoids-plant-material) Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS. - [See All (9)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search) ## How Can We Help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/CA/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional Support - [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search) Use the Chromatogram Search to identify unknown compounds in your sample. - [Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer Support Request](https://www.sigmaaldrich.com/CA/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services. * * * __Featured Articles__ [Comparison of QuEChERS Products for the Extraction of Planar Pesticides from Spinach](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/planar-pesticides-spinach) Supel™ QuE Verde combines a novel carbon with zirconia coated silica (Z-Sep+) to provide an optimum balance between planar pesticide recovery and color removal. [Pesticide Residue Analysis in Ginger Powder with QuEChERS](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/analysis-of-pesticide-residues-in-ginger-powder) 2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis. [Supel™ QuE Z-Sep Sorbents for QuEChERS](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/quechers) Z-Sep sorbents improve fat and pigment extraction in QuEChERs solid phase extraction for sample preparation. [Analysis of Pesticide Residues in Pistachios Using QuEChERS Extraction and Cleanup with Supel™ QuE Z-Sep+](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/analytical-chemistry/quechers/analysis-of-pesticide-residues-in-pistachios) Pistachios 45% fat complicates pesticide analysis. But Z-Sep+ sorbent cleanup after QuEChERS extraction can reduce background for reliable results. Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/CA/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/CA/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/CA/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/CA/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/CA/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/CA/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/CA/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/CA/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/CA/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/CA/en/collections/press)[Programs](https://www.sigmaaldrich.com/CA/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/CA/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/milliporesigma) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/MilliporeSigma) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/MilliporeSigma) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/milliporesigma/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@milliporesigma2407/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/CA/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/CA/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/CA/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/CA/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/CA/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/CA/en/site-map) An unknown error has occured.