[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/CA/en)
Products
Cart0
CAEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/CA/en)[Applications](https://www.sigmaaldrich.com/CA/en/applications)[Materials Science and Engineering](https://www.sigmaaldrich.com/CA/en/applications/materials-science-and-engineering) Electron Microscopy
# Electron Microscopy
 image and selected area electron diffraction (SAED) of Fe3O4 nanoparticles with the shape of tetrahedrons.")
Electron microscopy is a technique used to obtain ultrahigh resolution images of individual atoms of materials and internal structures of cells. The resulting atomic-level or micro- and meso-structure images can be used to investigate the sample properties and behavior. It is used in materials science, biomedical research, quality control, and failure analysis. The use of electrons as the imaging radiation source allows for greater spatial resolution (on the tens of picometers scale) as compared to the resolution achieved using photons in optical microscopy (~200 nanometers). In addition to surface topography, information about crystalline structure, chemical composition, and electrical properties are obtainable through electron microscopy. Electron microscopy can be divided into two main categories: scanning electron microscopy (SEM) and transmission electron microscopy (TEM).
* * *
## Related Products
Slide 1 of 20
1 of 5
[
\
Sigma-Aldrich
\
TEM-CF200CU50
\
Carbon Film Supported Copper Grid](https://www.sigmaaldrich.com/CA/en/product/aldrich/temcf200cu50)
Quick View
[
\
Sigma-Aldrich
\
TEM-7435703
\
Carbon Film Supported Copper Tomography Grid R14C2](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem7435703)
Quick View
[
\
Sigma-Aldrich
\
TEM-74357
\
Copper Tomography Grid R14C2](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem74357)
Quick View
[
\
Sigma-Aldrich
\
TEM-FCF400CU
\
Formvar/Carbon Supported Copper Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temfcf400cu)
Quick View
[
\
Sigma-Aldrich
\
TEM-FCF300AU
\
Formvar/Carbon Supported Gold Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temfcf300au)
Quick View
[
\
Sigma-Aldrich
\
TEM-FCF100NI
\
Formvar/Carbon Supported Nickel Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temfcf100ni)
Quick View
[
\
Sigma-Aldrich
\
TEM-HC300CUCC
\
Holey Carbon/Continuous Ultrathin carbon film Supported Copper Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temhc300cucc)
Quick View
[
\
Sigma-Aldrich
\
TEM-HC300CU100
\
Holey Carbon Supported Copper Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temhc300cu100)
Quick View
[
\
Sigma-Aldrich
\
TEM-LC200CU100
\
Lacey Carbon Supported Copper Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temlc200cu100)
Quick View
[
\
Sigma-Aldrich
\
TEM-LC400CU150
\
Lacey Carbon Supported Copper Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temlc400cu150)
Quick View
[
\
Sigma-Aldrich
\
TEM-7435701
\
Lacey Carbon Supported Copper Tomography Grid R14C2](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem7435701)
Quick View
[
\
Sigma-Aldrich
\
TEM-LC300AU
\
Lacey Carbon Supported Gold Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temlc300au)
Quick View
[
\
Sigma-Aldrich
\
TEM-LC200NI
\
Lacey Carbon Supported Nickel Grids](https://www.sigmaaldrich.com/CA/en/product/aldrich/temlc200ni)
Quick View
[
\
Sigma-Aldrich
\
TEM-7596407
\
OMNI Probe Copper 3-Post Lift-Out Grid](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem7596407)
Quick View
[
\
Sigma-Aldrich
\
TEM-7596410
\
OMNI Probe Copper 4-Post Lift-Out Grid](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem7596410)
Quick View
[
\
Sigma-Aldrich
\
TEM-7596406
\
OMNI Probe Molybdenum 4-Post Lift-Out Grid](https://www.sigmaaldrich.com/CA/en/product/aldrich/tem7596406)
Quick View
[
\
Sigma-Aldrich
\
G5882
\
Glutaraldehyde solution](https://www.sigmaaldrich.com/CA/en/product/sial/g5882)
Quick View
[
\
Sigma-Aldrich
\
G6257
\
Glutaraldehyde solution](https://www.sigmaaldrich.com/CA/en/product/sial/g6257)
Quick View
[
\
Sigma-Aldrich
\
74378
\
(2-Nonen-1-yl)succinic anhydride](https://www.sigmaaldrich.com/CA/en/product/sigma/74378)
Quick View
[
\
G4901
\
Grids for transmission electron microscopy](https://www.sigmaaldrich.com/CA/en/product/sigma/g4901)
Quick View
* * *
## Featured Categories
[](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/battery-materials)
[Battery Materials](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/battery-materials)
Battery materials ensure reproducible data, supporting research needs from bench-scale to manufacturing for reliable performance.
[Shop Products](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/battery-materials)
[](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/carbon-nanomaterials)
[Carbon Nanomaterials](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/carbon-nanomaterials)
Discover diverse carbon nanomaterials: fullerene, nanotubes, graphene, quantum dots, nanodiamonds. Fuel energy, electronics, therapeutics research.
[Shop Products](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/carbon-nanomaterials)
[](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/quantum-dots)
[Quantam Dots](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/quantum-dots)
Light up your research with our comprehensive portfolio of core-type, core-shell, and alloyed quantum dots in various compositions, sizes, functionalizations, and kits.
[Shop Products](https://www.sigmaaldrich.com/CA/en/products/materials-science/energy-materials/quantum-dots)
[](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
[Inorganic & Metallic Nanomaterials](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
We offer a comprehensive portfolio of inorganic and metallic nanomaterials, functionalized nanoparticles, and nanomaterial kits for your research needs.
[Shop Products](https://www.sigmaaldrich.com/CA/en/products/materials-science/electronic-materials/inorganic-and-metallic-nanomaterials)
* * *
Overview
Related Articles & Protocols
Support
Scanning electron microscopy (SEM) uses a relatively low-power electron beam for imaging and interaction with the sample. Electron detectors identify secondary electrons at the surface and backscattered electrons in deeper regions. Secondary electrons are generated from inelastic interactions between the electron beam and the atoms of the sample. Backscattered electrons are generated after elastic interaction between the electron beam and the sample. SEM requires little to no sample preparation and is much faster and less restrictive than other types of electron microscopy. Large (~200 millimeter) samples can be directly imaged after mounting to a holder or stub. SEM commonly uses energy-dispersive X-ray spectroscopy (EDS or EDX) to map the distribution of elements within a sample. Electron beam-induced current (EBIC) and cathodoluminescence (CL) are other methods to analyze the high-quality images and optoelectronic properties of samples.
Transmission electron microscopy (TEM) uses a high-energy beam of electrons to transmit electrons through a sample to create a 2D image at the highest possible resolution. Nanomaterials can be analyzed through TEM to reveal their structure and composition information at the atomic level. Selecting the right sample holder (TEM grid) for different types of nanomaterials is vital to obtain the most detailed information. When samples are too thick, they must first be made thin enough for electrons to travel through them, ideally 100 nanometers or less. These TEM samples are then mounted onto a TEM grid and studied under ultrahigh vacuum conditions with a focused, intense electron beam. TEM utilizes the selected area diffraction (SAD) of electrons passing through the sample to provide crystallographic information about the sample material. Electron energy loss spectroscopy (EELS) and energy-dispersive X-ray spectroscopy (EDX) are methods of analysis to measure the atomic composition, chemical bonding, electronic properties, and local material thickness.
Scanning transmission electron microscopy (STEM) scans a focused electron beam (with typical spot size of 0.05-0.2 nm) over the sample to complete imaging and spectroscopic mapping simultaneously, allowing direct correlation of spatial information and spectroscopic data.
[](https://www.sigmaaldrich.com/documents-search)
[Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search)
Visit our document search for data sheets, certificates and technical documentation.
[Find Documents](https://www.sigmaaldrich.com/documents-search)
## Related Articles
- [Atomic Layer Deposition of Nanomaterials for Li-Ion Batteries, Fuel Cells, and Solar Cells](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/atomic-layer-deposition-of-nanomaterials-for-li-ion-batteries-fu)
Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.
- [Nanomaterials for Energy Storage in Lithium-ion Battery Applications](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanomaterials-for0)
Nanomaterials for Energy Storage in Lithium-ion Battery Applications
- [Nano Olivine-based Cathode Materials for Li-ion Batteries](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanostructured-olivine-based-cathode-materials)
Researchers urged to develop renewable energy sources like wind and solar to mitigate fossil fuel's environmental impact.
- [Scaling Up High-Energy Cathode Materials for Electric Vehicles](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/scaling-up-high-energy-cathode-materials)
The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.
- [Fluorescent Nanodiamond Particles: Properties and Applications](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/fluorescent-nanodiamond-particles)
Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.
- [See All (30)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search)
## Related Protocols
- [Single-walled Carbon Nanotubes: Exfoliation and Debundling Procedure](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/materials-science-and-engineering/microelectronics-and-nanoelectronics/single-walled-carbon-nanotubes-exfoliation-debundling-procedure)
Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.
- [See All (1)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search)
### Find More Articles and Protocols
Enter Keywords
Search
## How Can We Help
In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/CA/en/support/customer-support)
or talk to our customer service team:
Email [custserv@sial.com](mailto:custserv@sial.com)
or call +1 (800) 244-1173
## Additional Support
- [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search)
Use the Chromatogram Search to identify unknown compounds in your sample.
- [Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps)
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- [Customer Support Request](https://www.sigmaaldrich.com/CA/en/support/customer-support)
Customer support including help with orders, products, accounts, and website technical issues.
- [FAQ](https://maestro.my.site.com/knowledgeportal/s/)
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.
* * *
__Featured Articles__
[Fluorescent Nanodiamond Particles: Properties and Applications](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/fluorescent-nanodiamond-particles)
Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.
[Nanomaterials for Energy Storage in Lithium-ion Battery Applications](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanomaterials-for0)
Nanomaterials for Energy Storage in Lithium-ion Battery Applications
[Nano Olivine-based Cathode Materials for Li-ion Batteries](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanostructured-olivine-based-cathode-materials)
Researchers urged to develop renewable energy sources like wind and solar to mitigate fossil fuel's environmental impact.
[Scaling Up High-Energy Cathode Materials for Electric Vehicles](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/scaling-up-high-energy-cathode-materials)
The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/CA/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/CA/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/CA/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/CA/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/CA/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/CA/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/CA/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/CA/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/CA/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/CA/en/collections/press)[Programs](https://www.sigmaaldrich.com/CA/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/CA/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/milliporesigma)
[](https://x.com/MilliporeSigma)
[](https://www.facebook.com/MilliporeSigma)
[](https://www.instagram.com/milliporesigma/)
[](https://www.youtube.com/@milliporesigma2407/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/CA/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/CA/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/CA/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/CA/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/CA/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/CA/en/site-map)
An unknown error has occured.