[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/CA/en) Products Cart0 CAEN Products [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/CA/en)[Applications](https://www.sigmaaldrich.com/CA/en/applications)[Microbiological Testing](https://www.sigmaaldrich.com/CA/en/applications/microbiological-testing)Bioburden Testing # Bioburden Testing ![Female scientist examining microbiological culture in petri dish.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/microbiological-testing/microbial-bioburden-and-water-testing.jpg "Microbial bioburden and water testing") Bioburden is the presence of viable microorganisms on a surface (or complete item), inside a device, or in a portion of liquid, before sterilization. Bioburden can be introduced from the raw materials used in the manufacturing process or through the workforce in the manufacturing environment or during the packaging of end products.  With numerous sources of potential contamination, the bioburden of a product can fluctuate between batches hence routine testing is implemented as a part of quality control.   * * * ## Related Products Slide 1 of 14 1 of 4 [![Milliflex Oasis® Filtration Unit pore size 0.45 μm, mixed cellulose esters (MCE) membrane, white filter, volume 100 mL](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/740/999/57d1a874-3e8b-4456-ba0b-9bea13c3b597/640/57d1a874-3e8b-4456-ba0b-9bea13c3b597.jpg) \ Millipore \ MMHAWG124 \ Milliflex Oasis® Filtration Unit](https://www.sigmaaldrich.com/CA/en/product/mm/mmhawg124) Quick View [![Milliflex® Quantum Standard Kit with validation protocol A4 format, For the rapid detection of microbial contamination](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/972/533/f9a807e1-1ae9-48f2-aff5-af69cf99d912/640/f9a807e1-1ae9-48f2-aff5-af69cf99d912.jpg) \ Millipore \ MXQUA4K01 \ Milliflex® Quantum Standard Kit](https://www.sigmaaldrich.com/CA/en/product/mm/mxqua4k01) Quick View [![Milliflex Oasis® Culture Media Plate suitable for (Tryptic soy media plates for aerobic and anaerobic microflora)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/340/014/d03ccec6-f3e3-45f0-b57e-e2355bf6b10d/640/d03ccec6-f3e3-45f0-b57e-e2355bf6b10d.jpg) \ Millipore \ MMSMCTS48 \ Milliflex Oasis® Culture Media Plate](https://www.sigmaaldrich.com/CA/en/product/mm/mmsmcts48) Quick View [![Milliflex Oasis® Culture Media Plate ready-to-use](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/413/705/c83bfc3d-aa07-436a-bcf6-e240dbf40c3b/640/c83bfc3d-aa07-436a-bcf6-e240dbf40c3b.jpg) \ Millipore \ MMSMCSD48 \ Milliflex Oasis® Culture Media Plate](https://www.sigmaaldrich.com/CA/en/product/mm/mmsmcsd48) Quick View [![Milliflex Oasis® Culture Media Plate suitable for (R2A low nutrient media plates for waterborne microorganisms)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/172/915/f3689899-b415-4685-aab9-66b7723cafef/640/f3689899-b415-4685-aab9-66b7723cafef.jpg) \ Millipore \ MMSMCRA48 \ Milliflex Oasis® Culture Media Plate](https://www.sigmaaldrich.com/CA/en/product/mm/mmsmcra48) Quick View [![EZ-Stream® vacuum pump suitable for bioburden testing, for use with EZ-Fit® Filtration Manifold](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/317/602/37cc1a9f-7c02-4f7d-8737-d390b740587b/640/37cc1a9f-7c02-4f7d-8737-d390b740587b.jpg) \ Millipore \ EZSTREAM1 \ EZ-Stream® vacuum pump](https://www.sigmaaldrich.com/CA/en/product/mm/ezstream1) Quick View [![EZ-Fit® Manifold 6-place, For EZ-Fit® Filtration Units](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/165/205/0d45b425-2e69-4da5-b4a0-798eb9c756f1/640/0d45b425-2e69-4da5-b4a0-798eb9c756f1.jpg) \ Millipore \ EZFITEFUN6 \ EZ-Fit® Manifold](https://www.sigmaaldrich.com/CA/en/product/mm/ezfitefun6) Quick View [![EZ-Fit® Manifold 3-place, For EZ-Fit® Filtration units](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/111/632/b0ba7611-c6c4-486f-81ec-ddf9b5596461/640/b0ba7611-c6c4-486f-81ec-ddf9b5596461.jpg) \ Millipore \ EZFITEFUN3 \ EZ-Fit® Manifold](https://www.sigmaaldrich.com/CA/en/product/mm/ezfitefun3) Quick View [![55-Plus™ monitor ready-to-use, disposable filtration unit, pore size 0.45 μm, white filter (gridded), mixed cellulose esters (MCE) membrane, suitable for (microbiological analysis), suitable for water monitoring](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/344/304/25f962b0-cb57-49a5-b4f7-2a389a2b50c4/640/25f962b0-cb57-49a5-b4f7-2a389a2b50c4.jpg) \ Millipore \ JBRMHWG05505 \ 55-Plus™ monitor](https://www.sigmaaldrich.com/CA/en/product/mm/jbrmhwg05505) Quick View [![Tryptone Glucose Extract Broth ampoule capacity 2 mL, ampoule filling volume 2 mL, ready-to-use, pack of 50 tests, for heterotrophic bacteria](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/313/178/88cc8756-5e09-462b-99fb-cf877e6d5018/640/88cc8756-5e09-462b-99fb-cf877e6d5018.jpg) \ Millipore \ MHA000P2T \ Tryptone Glucose Extract Broth](https://www.sigmaaldrich.com/CA/en/product/mm/mha000p2t) Quick View [![Tryptic Soy Agar - Dehydrated Culture Media GranuCult® prime, EP, USP, JP, ISO 11133, FDA (BAM), granular, suitable for microbiology](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/295/209/27e42456-0eb4-4281-8870-c5b50448ab81/640/27e42456-0eb4-4281-8870-c5b50448ab81.jpg) \ Millipore \ 1.05458 \ Tryptic Soy Agar - Dehydrated Culture Media](https://www.sigmaaldrich.com/CA/en/product/mm/105458) Quick View [![Tryptic Soy Agar - Ready-to-use Settle Plates ReadyPlate™, plate diam. 90 mm, according to ISO, FDA-BAM, EP, USP, sterile, pack of 20 and 100 plates](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/355/786/ff04018b-3adf-4fa0-b833-b9c4d1e28600/640/ff04018b-3adf-4fa0-b833-b9c4d1e28600.jpg) \ Millipore \ 1.46431 \ Tryptic Soy Agar - Ready-to-use Settle Plates](https://www.sigmaaldrich.com/CA/en/product/mm/146431) Quick View [![Plate Count Agar ReadyPlate™, diam. 55 mm, ready-to-use, sterile, box of 20 plates, for aerobic bacteria](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/657/351/1617f536-2a54-4fde-83e7-43005954d03f/640/1617f536-2a54-4fde-83e7-43005954d03f.jpg) \ Millipore \ 1.46763 \ Plate Count Agar](https://www.sigmaaldrich.com/CA/en/product/mm/146763) Quick View [![R2A Agar Chromogenic Heimplate, plate diam. 90 mm, ready-to-use, settle Plate for long Incubation, aseptically filled, box of 20 plates, for heterotrophic bacteria](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/313/483/cf9ab428-d7bc-4919-8edb-bb1226f56e33/640/cf9ab428-d7bc-4919-8edb-bb1226f56e33.jpg) \ Millipore \ 1.46049 \ R2A Agar Chromogenic](https://www.sigmaaldrich.com/CA/en/product/mm/146049) Quick View * * * ## Featured Categories [![Microbial Filtration](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/microbial-filtration/ez-product-family.jpg "EZ Product Family for membrane filtration")](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-filtration) [Microbial Filtration](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-filtration) Speed up your time-to-result with rapid microbial testing using our complete membrane filtration systems; used in bioburden testing, food and beverage analysis, and cannabis microbiology testing. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-filtration) [![Microbial Culture Media](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/granucult-prime-macconkey-agar-culture-media-granules.jpg "GranuCult® Prime MacConkey Agar for Microbiology")](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) [Microbial Culture Media](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) Discover high-quality microbial culture media. Choose from dehydrated or ready-to-use options, meeting industry standards and regulatory requirements. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) [![Milli-Q® Benchtop Lab Water Purification Systems](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/water-purification/ziq7003t0c-utrapure-pure-water-purification-system64.jpg "Milli-Q® IQ 7003/05/10/15 Pure and Ultrapure Water Purification System 64")](https://www.sigmaaldrich.com/CA/en/products/water-purification/milli-q-benchtop-water-purification-systems) [Milli-Q® Benchtop Lab Water Purification Systems](https://www.sigmaaldrich.com/CA/en/products/water-purification/milli-q-benchtop-water-purification-systems) Milli-Q® systems offer innovative water purification technologies engineered to support lab research needs, sustainability goals, and other major requirements. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/water-purification/milli-q-benchtop-water-purification-systems) * * * Overview Related Articles & Protocols Support ## Fundamentals of Bioburden testing Bioburden testing or Microbial testing is a quality control process that detects and quantifies microbial contamination of a product at different stages of production i.e., from initial manufacturing to final distribution. Effective quality control and accurate test results are essential to minimize risks for consumers and are required by regulated production environments. Therefore, bioburden analysis is often included in routine testing to ensure the safety, quality, and regulatory compliance of each manufactured product batch. Bioburden testing is performed for medical devices, pharmaceuticals, food and beverages, water, packaging, raw materials, human tissue, animal tissue, and cosmetics. When standard methods, as described below are followed, it is important to ensure that the testing method does not introduce bacteria into the test sample or kill bacteria in the test sample. - [Membrane Filtration Method for Bioburden Testing](https://www.sigmaaldrich.com#membrane) - [Direct Plating Methods for Bioburden Testing](https://www.sigmaaldrich.com#direct) - [Most Probable Number (MPN) Method for Bioburden Testing](https://www.sigmaaldrich.com#most) [Request Information](https://www.sigmaaldrich.com/CA/en/campaigns/microbial-filtration) ## [](https://www.sigmaaldrich.com)Membrane Filtration Method for Bioburden Testing [Membrane filtration](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-filtration) is the method of choice for products containing antimicrobial substances. In this method, the sample is passed through a membrane filter with a 0.45 µm pore size. The membrane functions as a barrier and captures microorganisms larger than the membrane pore size. During filtration, a vacuum can be used to speed up the process. The membrane is then transferred to a culture medium and placed in an incubator for at least 5 days at 30–35 °C for bacterial detection and at 20-25 °C for fungal detection. The resulting culture is further enumerated to determine the levels of microbial contamination in the sample. Appropriate measures should be taken to avoid cross-contamination that could lead to false-positive results. ## [](https://www.sigmaaldrich.com)Direct Plating Methods For Bioburden Testing Direct plating methods for bioburden testing include pour plate and spread plate methods. The pour plate method is preferred due to its higher theoretical accuracy. In the pour plate method, the sterilized [culture medium](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) is added to a petri dish containing the test sample and allowed to solidify. Conversely, in the spread plate method, the sample is added to a petri dish containing sterile, solidified culture media. Regardless of the method, after the sample has been added to the culture system, the plate is incubated, and the resulting culture is enumerated. ## [](https://www.sigmaaldrich.com)Most Probable Number (MPN) Method For Bioburden Testing The most probable number (MPN) method is a quantitative method used to determine the approximate bacterial concentration in a sample. The original solution or sample is subdivided by orders of magnitude (frequently 10× or 2×) into culture broth and assessed for the presence or absence of microorganisms. The drawback of the MPN method is that it requires large numbers of replicates at the appropriate dilution to narrow the confidence intervals. Furthermore, the method is only effective for bacterial examination and does not provide reliable results for the enumeration of fungi. [![Document Search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png "Document Search")](https://www.sigmaaldrich.com/documents-search) [Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find Documents](https://www.sigmaaldrich.com/documents-search) ## Related Articles - [Fluorescence-based Rapid Microbial Detection](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/milliflex-quantum-rapid-detection) Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology. - [Comparison of Single-Use Funnels vs. Reusable Funnels for the Membrane Filtration Method in QC Microbiology](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/single-use-funnels-vs-reusable-funnels-membrane-filtration-method-qc-microbiology) Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology. - [S-Pak® Membrane Filters](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/s-pak-membrane-filter) S-Pak® membrane filters for microbial bioburden testing - [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media) Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms - [EN ISO 11133 Microbiological Culture Media](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/en-iso-11133-water-for-the-preparation-and-performance-testing) Water source comparison for microbiological media preparation highlights central purification vs. Milli-Q® IX system efficiency. - [See All (10)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related Protocols - [Routine Water and Beverage Microbiological Analysis Using the 55-Plus™ Monitor](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/microbiological-testing/bioburden-testing/55-plus-monitor) Routine water and beverage microbiological analysis using the 55-Plus™ Monitor - [MicropreSure® On-line Filtration System for In-process Microbiology Testing](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/microbiological-testing/bioburden-testing/micropresure-filtration) MicropreSure® on-line filtration system for in-process microbiology testing - [Microbial Growth Protocols](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/microbial-cell-culture/microbial-growth) General protocols for growth of competent cells in microbial medium. ### Find More Articles and Protocols Enter Keywords Search ## How Can We Help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/CA/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional Support - [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search) Use the Chromatogram Search to identify unknown compounds in your sample. - [Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer Support Request](https://www.sigmaaldrich.com/CA/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services. [![Showcases laboratory equipment, including a clear container being filled with a blue liquid from a flask held by a person wearing a purple glove. The background features vibrant pink and yellow colors, with the Millipore logo displayed in the lower right corner, emphasizing the brand's identity in the scientific field.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/banners/promotional/milliflex-oasis-cross-promo/milliflex-oasis-cross-promo-mobile-hero-290x217.jpg) \ Increase Result Reliability \ The touch-free membrane transfer technology of the Milliflex Oasis® system ensures consistent results from every member of your team. \ Learn More](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/milliflex-oasis-filtration) * * * __Featured Articles__ [Fluorescence-based Rapid Microbial Detection](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/milliflex-quantum-rapid-detection) Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology. [Comparison of Single-Use Funnels vs. Reusable Funnels for the Membrane Filtration Method in QC Microbiology](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/single-use-funnels-vs-reusable-funnels-membrane-filtration-method-qc-microbiology) Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology. [S-Pak® Membrane Filters](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/bioburden-testing/s-pak-membrane-filter) S-Pak® membrane filters for microbial bioburden testing [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media) Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register An unknown error has occured. - English - EN - Français - FR [Learn More](https://www.sigmaaldrich.com)