[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/CA/en) Products Cart0 CAEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/CA/en)[Applications](https://www.sigmaaldrich.com/CA/en/applications)[Microbiological Testing](https://www.sigmaaldrich.com/CA/en/applications/microbiological-testing)Pathogen & Spoilage Testing # Pathogen & Spoilage Testing ![Salmonella pathogenic rod-shaped bacteria.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/microbiological-testing/pathogenic-bacteria-salmonella.jpg "Pathogenic bacteria Salmonella") Pathogen testing is used to identify pathogenic organisms in manufacturing environments, ingredients or finished products that could harm the consumer. Types of pathogens include specific bacteria strains, viruses, fungi, or parasites that can cause illness in its host. Pathogen testing is continuously performed throughout the production process from raw materials to finished product screening to ensure consumer safety and is subject to strict food safety regulatory guidelines. * * * ## Related Products Slide 1 of 16 1 of 4 [![GDS Salmonella Tq](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/357/012/3426cdaf-f78a-4d1f-aa9e-32bcaf366bb5/640/3426cdaf-f78a-4d1f-aa9e-32bcaf366bb5.jpg) \ Millipore \ 71008BC \ GDS *Salmonella* Tq](https://www.sigmaaldrich.com/CA/en/product/sial/71008bc) Quick View [![GDS E. coli O157:H7 Tq BioControl, Molecular based PCR test for the detection of E. coli O157:H7 in food and environmental samples](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/847/787/673dea38-8a51-4b8a-a757-ae0b08dc0299/640/673dea38-8a51-4b8a-a757-ae0b08dc0299.jpg) \ Millipore \ 71007BC \ GDS *E. coli* O157:H7 Tq](https://www.sigmaaldrich.com/CA/en/product/sial/71007bc) Quick View [![MC-Media Pad® RAC for aerobic bacteria, ready-to-use, pack of 4 × 25 pieces](https://www.sigmaaldrich.com/assets/images/millipore-no-image/millipore-no-image_w640.png) \ Millipore \ 1.32359 \ MC-Media Pad® RAC](https://www.sigmaaldrich.com/CA/en/product/mm/132359) Quick View [![MC-Media Pad® EC for coliforms, for Escherichia coli, ready-to-use, pack of 4 × 25 pieces](https://www.sigmaaldrich.com/assets/images/millipore-no-image/millipore-no-image_w640.png) \ Millipore \ 1.32357 \ MC-Media Pad® EC](https://www.sigmaaldrich.com/CA/en/product/mm/132357) Quick View [![SimPlate® Yeast & Mold BioControl, Quantitative results for yeast and mold from food and environmental samples](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/146/643/9d810add-d3ae-4424-95ba-835d3164d136/640/9d810add-d3ae-4424-95ba-835d3164d136.jpg) \ Millipore \ 66004BC \ SimPlate® Yeast & Mold](https://www.sigmaaldrich.com/CA/en/product/sial/66004bc) Quick View [![VIP® Gold Listeria BioControl, Lateral flow test for detection of Listeria in food and environmental samples](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/420/648/8172045d-f714-41cc-b91b-6cf2580ee64e/640/8172045d-f714-41cc-b91b-6cf2580ee64e.jpg) \ Millipore \ 60037BC \ VIP® Gold *Listeria*](https://www.sigmaaldrich.com/CA/en/product/sial/60037bc) Quick View [![TRANSIA® AG Salmonella BioControl, ELISA test for detection of Salmonella in food and environmental samples](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/157/075/49207506-6191-48f2-b228-4adeec3a4acd/640/49207506-6191-48f2-b228-4adeec3a4acd.jpg) \ Millipore \ 61001BC \ TRANSIA® AG *Salmonella*](https://www.sigmaaldrich.com/CA/en/product/sial/61001bc) Quick View [![DiluCult™ Gravimetric Dilutor, suitable for sample preparation](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/132/121/02ddf735-4116-49ee-9029-5dd50207dd65/640/02ddf735-4116-49ee-9029-5dd50207dd65.jpg) \ Millipore \ 5.42753 \ DiluCult™](https://www.sigmaaldrich.com/CA/en/product/mm/542753) Quick View [![Enrichment Sample Homogenizer (ESH) sample homogenizer, suitable for sample preparation](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/280/096/90841075-2b60-46c6-836a-c719a581806a/640/90841075-2b60-46c6-836a-c719a581806a.jpg) \ Millipore \ 5.42765 \ Enrichment Sample Homogenizer (ESH)](https://www.sigmaaldrich.com/CA/en/product/mm/542765) Quick View [![Buffered Peptone Water ReadyTube®, ISO 6887, ISO 6579, bottle capacity 1 L, bottle filling volume 1 L, closure type, Blue screw cap with 3 loci, box of 6 bottles](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/149/218/4685c720-8e34-4a23-bd2d-daa4d27ba444/640/4685c720-8e34-4a23-bd2d-daa4d27ba444.jpg) \ Millipore \ 1.46403 \ Buffered Peptone Water](https://www.sigmaaldrich.com/CA/en/product/mm/146403) Quick View [![Tryptic Soy Agar - Ready-to-use Settle Plates ReadyPlate™, plate diam. 90 mm, according to ISO, FDA-BAM, EP, USP, sterile, pack of 20 and 100 plates](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/355/786/ff04018b-3adf-4fa0-b833-b9c4d1e28600/640/ff04018b-3adf-4fa0-b833-b9c4d1e28600.jpg) \ Millipore \ 1.46431 \ Tryptic Soy Agar - Ready-to-use Settle Plates](https://www.sigmaaldrich.com/CA/en/product/mm/146431) Quick View [![Buffered Peptone Water GranuCult® prime, ISO 19250, ISO 6579-1, ISO 21528-1, ISO 22964, FDA-BAM, EP, granular, pkg of 0.5-25 kg](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/421/938/d5697a73-a483-4db7-ad3d-f9182cb996b2/640/d5697a73-a483-4db7-ad3d-f9182cb996b2.jpg) \ Millipore \ 1.07228 \ Buffered Peptone Water](https://www.sigmaaldrich.com/CA/en/product/mm/107228) Quick View [![Plate Count agar GranuCult® prime, ISO 4833, IFU, ISO 17410, FDA-BAM, granular, suitable for food and beverages](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/251/110/c1383e21-a0ad-434c-ad58-d2f1b1d1e1c1/640/c1383e21-a0ad-434c-ad58-d2f1b1d1e1c1.jpg) \ Millipore \ 1.05463 \ Plate Count agar](https://www.sigmaaldrich.com/CA/en/product/mm/105463) Quick View [![Singlepath® Campylobacter suitable for lateral flow assay, for Campylobacter spp., input: food(s)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/643/446/87d55a5e-a139-4c23-84ff-c9f9d5848df8/640/87d55a5e-a139-4c23-84ff-c9f9d5848df8.jpg) \ Millipore \ 1.04143 \ Singlepath® *Campylobacter*](https://www.sigmaaldrich.com/CA/en/product/mm/104143) Quick View [![Singlepath® Listeria monocytogenes suitable for lateral flow assay, for Listeria monocytogenes, input: food(s)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/283/407/8469187a-7134-439a-bfeb-9877a5a580b1/640/8469187a-7134-439a-bfeb-9877a5a580b1.jpg) \ Millipore \ 1.04148 \ Singlepath® *Listeria monocytogenes*](https://www.sigmaaldrich.com/CA/en/product/mm/104148) Quick View [![Readybag® Half Fraser Broth with Supplements acc. ISO 11290, 62 g, irradiated, For the primary selective enrichment of Listeria sp.](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/713/243/30c13f4e-5cec-415e-951f-371bdebe989e/640/30c13f4e-5cec-415e-951f-371bdebe989e.jpg) \ Millipore \ 1.01865 \ Readybag® Half Fraser Broth with Supplements](https://www.sigmaaldrich.com/CA/en/product/mm/101865) Quick View * * * ## Featured Categories [![Bottle with piles of microbial culture media](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/microbial-culture-media-mcp.jpg)](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) [Microbial Culture Media](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) Discover high-quality microbial culture media. Choose from dehydrated or ready-to-use options, meeting industry standards and regulatory requirements. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media) [![Pathogen Screening Kits, Instruments & Disposables](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/rotor-gene-operator-tube.jpg)](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/pathogen-screening-kits-instuments-and-disposables) [Pathogen Screening Kits, Instruments & Disposables](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/pathogen-screening-kits-instuments-and-disposables) Pathogen testing is a key component and critical success factor in effective food safety and process control programs. Explore our wide range of pathogen screening products that suit your needs. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/pathogen-screening-kits-instuments-and-disposables) [![Bottle with piles of microbial culture media raw materials](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/microbial-culture-media-rm.jpg)](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media-raw-materials) [Microbial Culture Media Raw Materials](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media-raw-materials) Explore our wide range of culture media raw materials and supplements for microbiological purposes. All our raw materials and culture media supplements are safe to handle and ensure optimal performance. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/microbial-culture-media-raw-materials) [![The inoculate step of the culture media methods workflow](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/industrial-microbiology/mc-media-pad-culture-media.jpg)](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/quality-indicator-testing-devices) [Quality Indicator Testing Devices](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/quality-indicator-testing-devices) Our collection of testing systems includes alternative & rapid culture media for fast & reliable indicator organisms testing results in a time-efficient method. [Shop Products](https://www.sigmaaldrich.com/CA/en/products/industrial-microbiology/quality-indicator-testing-devices) * * * Overview Related Articles & Protocols Support ![A person in protective clothing, including a hairnet and face mask, working in a laboratory with various bottles of chemicals on the countertop. Background with a clean, organized workspace, including a laptop and scientific equipment.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/microbiological-testing/pathogen-and-spoilage-testing-webpage-picture.jpg "Pathogen and Spoilage Testing Webpage Picture") ## [](https://www.sigmaaldrich.com)BIOSAFETY TESTING SERVICES [Biosafety testing services](https://www.sigmaaldrich.com/CA/en/campaigns/bioreliance-biosafety-testing-and-product-characterization) to investigate mAbs and cell and gene therapy and vaccines materials to ensure that they are free of adventitious agents or unexpected results that can cause catastrophic failures in downstream processing, including: Microbiology and Molecular testing. ## [](https://www.sigmaaldrich.com)Pathogen Detection Methods - Conventional cultivation-based enrichment and plating methods and rapid plating solution - Ready-to-use media methods - ELISA and immunoassay technology - PCR and molecular detection - Lateral flow tests ## [](https://www.sigmaaldrich.com)Indicator Organisms While pathogen testing is looking for specific pathogens, indicator organism testing is looking to verify overall sanitation and safety. Microbial indicators are microorganisms that are not necessarily pathogenic but indicate the potential presence of pathogens. Detecting indicator organisms means that the product has been exposed to conditions that could lead to an increased risk of pathogenic contamination. Pathogens can be difficult to detect. Therefore, indicator organism tests are often used as additional methods to assess the hygienic conditions of a product or manufacturing environment. ## [](https://www.sigmaaldrich.com)Microbial Spoilage Testing Microbial spoilage testing is the process of determining the presence of spoilage microorganisms that cause product deterioration, effecting the quality and safety. These include Gram-positive and Gram-negative bacteria, as well as yeasts and molds. Spoilage microorganisms are not pathogenic, but can change the physical properties, such as texture, smell, taste, and color, or chemical properties of a product and making it unsuitable for consumption. Spoilage can occur at any stage along the food chain and needs to be part of the microbiological quality control. The testing requirements vary from industry to industry, depending on different spoilage parameters. [![Document Search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png)](https://www.sigmaaldrich.com/documents-search) [Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find Documents](https://www.sigmaaldrich.com/documents-search) ## Related Articles - [Differentiation and Detection of Escherichia coli, Coliforms and Enterobacteriaceae](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/selective-growth-media) An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms. - [Microbiology Quality Control in Food and Beverage](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/microbio-qc-food-and-beverage) Food contaminated by microorganisms (bacteria and yeasts), viruses, and protozoa can cause severe disease. - [HybriScan® Rapid Microbial Test System](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/hybriscan-rapid-test) Fast molecular screening of beverage, water, wastewater and food for microorganisms - [*Salmonella* Contamination and Rapid Detection Methods](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/salmonella-contamination-and-testing) Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments. - [Detection of Listeria monocytogenes in Food and Environmental Samples](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/listeria-monocytogenes) Detection of Listeria monocytogenes in food and environmental samples according to ISO 11290-1, FDA-BAM and USDA-FSIS - [See All (43)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related Protocols - [Study: Comparison of MC-Media Pad® with Different Convenience Solution Products](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/microbiological-testing/microbial-culture-media-preparation/mc-media-pad-product-comparison) Comparison of MC-Media Pad® with different convenience solution products for indicator organism testing - [Yeast Growth Methods](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/microbiological-testing/pathogen-and-spoilage-testing/yeast-growth-protocols) Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth. - [Revised EN ISO 21528:2017 for Detection and Enumeration of *Enterobacteriaceae* in the Food Chain](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/food-and-beverage-testing-and-manufacturing/regulatory-compliance-for-food-and-beverage/iso-21528-enterobacteriaceae-detection-and-enumeration) Learn about ISO 21528 standards for detecting and enumerating Enterobacteriaceae in food and animal feed. Enhance safety and compliance with food production. - [*Salmonella* Detection in the Food Chain (EN ISO 6579-1)](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/food-and-beverage-testing-and-manufacturing/regulatory-compliance-for-food-and-beverage/iso-6579-1-salmonella-detection) EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples. - [Validation of Shorter Protocol for Detection of *Salmonella enterica* subsp. *enterica* in Peanut Butter Samples Followed by a rRNA Detection System](https://www.sigmaaldrich.com/CA/en/technical-documents/protocol/microbiological-testing/pathogen-and-spoilage-testing/protocol-for-detection-of-salmonella-enterica) Food-borne pathogens Salmonella is commonly evaluated in manufacturing of peanut butter and other food products. - [See All (11)](https://www.sigmaaldrich.com/CA/en/search/facet-search?focus=sitecontent&term=facet-search) ### Find More Articles and Protocols Enter Keywords Search ## How Can We Help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/CA/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional Support - [Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer Support Request](https://www.sigmaaldrich.com/CA/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services. * * * __Featured Articles__ [Differentiation and Detection of Escherichia coli, Coliforms and Enterobacteriaceae](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/selective-growth-media) An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms. [Molecular Detection for Rapid Food Pathogen Testing](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/assurance-gds-genetic-detection) Assurance® GDS is an integrated solution for food pathogen detection. It offers faster pathogen testing results with high accuracy to meet the demanding needs of food, environmental and cannabis microbiology testing challenges. [*Salmonella* Contamination and Rapid Detection Methods](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/salmonella-contamination-and-testing) Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments. [Anaerobic Culture with Anaerocult®](https://www.sigmaaldrich.com/CA/en/technical-documents/technical-article/microbiological-testing/pathogen-and-spoilage-testing/anaerobic-incubation-system) Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs. Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/CA/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/CA/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/CA/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/CA/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/CA/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/CA/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/CA/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/CA/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/CA/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/CA/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/CA/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/CA/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/CA/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/CA/en/collections/press)[Programs](https://www.sigmaaldrich.com/CA/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/CA/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/milliporesigma) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/MilliporeSigma) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/MilliporeSigma) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/milliporesigma/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@milliporesigma2407/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/CA/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/CA/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/CA/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/CA/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/CA/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/CA/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/CA/en/site-map)