AC5372908088 Accela ChemBio Inc.

2,6-Dinitrobenzonitrile

  • MDL number MFCD00041246
  • Cas Number : 35213-00-4
  • Molecular Weight : 193.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY002486&attname=GoodsCode

Safety Information

signalword Danger
pcodes P260-P261-P301+P310-P305+P351+P338-P405-P501A
hcodes GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY002486.pdf
hstatements H301-H312-H332-H315-H319-H373
Availability details for
Quantity
  • Availability
  • Add to Cart