AC5372908602 Accela ChemBio Inc.

2-(Methylsulfonamido)benzaldehyde

  • MDL number MFCD11052341
  • Cas Number : 94532-99-7
  • Molecular Weight : 199.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003552&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003552.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart