AC5372908608 Accela ChemBio Inc.

Methyl 4-Pyridinylacetate

  • MDL number MFCD00972014
  • Cas Number : 29800-89-3
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003565&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY003565.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart