AC5372908633 Accela ChemBio Inc.

4-(5-Oxazolyl)phenol

  • MDL number MFCD00207683
  • Cas Number : 1128-71-8
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003622&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003622.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart