AC5372908648 Accela ChemBio Inc.

5-Hydroxypiperidine-3-carboxylic Acid

  • MDL number MFCD02181125
  • Cas Number : 885951-97-3
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003650&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003650.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart