AC5372908716 Accela ChemBio Inc.

Ethyl Piperidin-4-ylacetate

  • MDL number MFCD02183575
  • Cas Number : 59184-90-6
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003777&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003777.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart