AC5372908760 Accela ChemBio Inc.

(1-Cbz-3-piperidine)carbothioamide

  • MDL number MFCD11223487
  • Cas Number : 569348-15-8
  • Molecular Weight : 278.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003855&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY003855.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart