AC5372908817 Accela ChemBio Inc.

4-Chloro-3-methylquinoline

  • MDL number MFCD09264040
  • Cas Number : 63136-60-7
  • Molecular Weight : 177.63022

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY003960&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY003960.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart