AC5372908836 Accela ChemBio Inc.

3-(Dimethylamino)-1-(4-methoxyphenyl)-propan-1-one Hydrochloride

  • MDL number MFCD00156682
  • Cas Number : 2125-49-7
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004000&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004000.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart