AC5372908841 Accela ChemBio Inc.

Methyl 4-(4-Fluorophenyl)-4-oxobutanoate

  • MDL number MFCD03090514
  • Cas Number : 39560-31-1
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004009&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004009.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart