AC5372908868 Accela ChemBio Inc.

1-Boc-5-Hydroxypiperidine-3-carboxylic Acid

  • MDL number MFCD11656771
  • Cas Number : 1095010-48-2
  • Molecular Weight : 245.27226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004056&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004056.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart