AC5372908897 Accela ChemBio Inc.

N-Boc-3-(aminomethyl)-3-methyloxetane

  • MDL number MFCD11656774
  • Cas Number : 208105-83-3
  • Molecular Weight : 201.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004115&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004115.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart