AC5372908918 Accela ChemBio Inc.

3-Methylsulfonylphenylacetic Acid

  • MDL number MFCD03426066
  • Cas Number : 1877-64-1
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004168&attname=GoodsCode

Safety Information

signalword Danger
pcodes P261-P280-P305 + P351 + P338
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY004168.pdf
hstatements H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart