AC5372908992 Accela ChemBio Inc.

1-(2-Cyanophenyl)piperidine-4-carboxylic Acid

  • MDL number MFCD08689748
  • Cas Number : 937601-79-1
  • Molecular Weight : 230.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004374&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004374.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart