AC5372908994 Accela ChemBio Inc.

Ethyl 2-(2-Cyanoanilino)acetate

  • MDL number MFCD00141059
  • Cas Number : 87223-76-5
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004376&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004376.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart