AC5372909145 Accela ChemBio Inc.

cis-5-Boc-1H-Hexahydropyrrolo[3,4-b]pyrrole

  • MDL number MFCD11045908
  • Cas Number : 180975-51-3
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004723&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004723.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart