AC5372909146 Accela ChemBio Inc.

4-(4-Methoxyphenyl)morpholine

  • MDL number MFCD00222002
  • Cas Number : 27347-14-4
  • Molecular Weight : 193.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004725&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004725.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart