AC5372909211 Accela ChemBio Inc.

2-Amino-4-(1-pyrazolyl)benzoic Acid

  • MDL number MFCD11656802
  • Cas Number : 1186663-55-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004876&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004876.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart