AC5372909212 Accela ChemBio Inc.

2-Amino-4-morpholinobenzoic Acid

  • MDL number MFCD11656803
  • Cas Number : 404010-74-8
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004877&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004877.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart