AC5372909225 Accela ChemBio Inc.

4-Benzyl-2-morpholinecarboxylic Acid Hydrochloride

  • MDL number MFCD02682031
  • Cas Number : 135072-15-0
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004904&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY004904.pdf
hstatements H302-H312-H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart