AC5372909238 Accela ChemBio Inc.

Methyl (S)-2-(N-Boc-Amino)-3-(5-methoxyindol-3-yl)propionate

  • MDL number MFCD11656820
  • Cas Number : 1235346-28-7
  • Molecular Weight : 348.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004925&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004925.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart