AC5372909331 Accela ChemBio Inc.

4-Benzyl-5-oxomorpholine-3-carboxylic Acid

  • MDL number MFCD11042914
  • Cas Number : 106910-79-6
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005112&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005112.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart