AC5372909361 Accela ChemBio Inc.

4-(Trifluoromethyl)thiazole-2-carboxylic Acid

  • MDL number MFCD07377490
  • Cas Number : 944900-55-4
  • Molecular Weight : 197.1350896

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005167&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005167.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart