AC5372909411 Accela ChemBio Inc.

2-Methyl-3-nitrothiophene-5-carboxylic Acid

  • MDL number MFCD00507024
  • Cas Number : 36050-35-8
  • Molecular Weight : 187.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005273&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005273.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart