AC5372909428 Accela ChemBio Inc.

3-Chloro-4-(methylsulfonyl)nitrobenzene

  • MDL number MFCD08692409
  • Cas Number : 91842-77-2
  • Molecular Weight : 235.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005306&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY005306.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart