AC5372909434 Accela ChemBio Inc.

6-Benzyl-6,7-dihydro-5H-pyrrolo[3,4-b]pyridine

  • MDL number MFCD06858493
  • Cas Number : 109966-30-5
  • Molecular Weight : 210.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005328&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005328.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart