AC5372909435 Accela ChemBio Inc.

Methyl Tetrahydropyran-2-carboxylate

  • MDL number MFCD12198853
  • Cas Number : 84355-44-2
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005329&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005329.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart