AC5372909479 Accela ChemBio Inc.

1-Acetyl-4-bromo-1H-indazole

  • MDL number MFCD12198857
  • Cas Number : 885698-70-4
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005435&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005435.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart