AC5372909481 Accela ChemBio Inc.

1-Hydroxy-7-methyllisoquinoline

  • MDL number MFCD12198859
  • Cas Number : 26829-47-0
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005440&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005440.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart