AC5372909495 Accela ChemBio Inc.

3-(1-Imidazolylmethyl)aniline

  • MDL number MFCD07801129
  • Cas Number : 120107-85-9
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005460&attname=GoodsCode
Availability details for
Quantity
  • Availability
  • Add to Cart