AC5372909497 Accela ChemBio Inc.

2-(Tetrahydropyran-4-yloxy)aniline

  • MDL number MFCD09025872
  • Cas Number : 898289-35-5
  • Molecular Weight : 193.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005464&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302 + P352-P261-P301+ P312- P304 + P340-P280-P305 + P351 + P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005464.pdf
hstatements H332-H302- H312-H315-H335-H319
Availability details for
Quantity
  • Availability
  • Add to Cart