AC5372909498 Accela ChemBio Inc.

Morpholino(2-piperidinyl)methanone Hydrochloride

  • MDL number MFCD05865124
  • Cas Number : 690634-79-8
  • Molecular Weight : 234.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005468&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005468.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart