AC5372909521 Accela ChemBio Inc.

2-(Aminomethyl)-3-phenylpropionic Acid

  • MDL number MFCD01549677
  • Cas Number : 95598-13-3
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005517&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005517.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart