AC5372909569 Accela ChemBio Inc.

4-Amino-1-(3-pyridyl)piperidine

  • MDL number MFCD09928096
  • Cas Number : 933760-08-8
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005626&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005626.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart