AC5372909577 Accela ChemBio Inc.

(R)-N-Acetyl-5-Chloro-Trp-OMe

  • MDL number MFCD12198866
  • Cas Number : 114872-81-0
  • Molecular Weight : 294.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005663&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005663.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart