AC5372909578 Accela ChemBio Inc.

(R)-N-Acetyl-6-Chloro-Trp-OMe

  • MDL number MFCD12198867
  • Cas Number : 1235280-35-9
  • Molecular Weight : 294.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005664&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005664.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart