AC5372909580 Accela ChemBio Inc.

(R)-N-Acetyl-6-Fluoro-Trp-OMe

  • MDL number MFCD12198869
  • Cas Number : 1234842-62-6
  • Molecular Weight : 278.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005666&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005666.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart