AC5372909597 Accela ChemBio Inc.

2,6-Dichloro-3-methylaniline

  • MDL number MFCD00007676
  • Cas Number : 64063-37-2
  • Molecular Weight : 176.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005706&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005706.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart