AC5372909600 Accela ChemBio Inc.

1-(6-Chloro-3-pyridazinyl)piperidin-4-ol

  • MDL number MFCD05864736
  • Cas Number : 89937-26-8
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005710&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005710.pdf
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart