AC5372909601 Accela ChemBio Inc.

3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoic Acid

  • MDL number MFCD08061016
  • Cas Number : 859155-81-0
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005714&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005714.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart