AC5372909634 Accela ChemBio Inc.

Methyl N-Boc-3-Oxopiperidine-4-carboxylate

  • MDL number MFCD12198882
  • Cas Number : 220223-46-1
  • Molecular Weight : 257.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005800&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005800.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart